Product Details
- SNP ID
-
rs13374837
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
56
- Location
-
Chr.1:21813855 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCAGGAAGAAAAGGGGCTCCTTA[C/T]CACACAGCTCTCCCCCTCCCCTTTT
- Phenotype
-
MIM: 142461
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSPG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79500108] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSPG2
- Gene Name
- heparan sulfate proteoglycan 2
There are no transcripts associated with this gene.
- Gene
- LDLRAD2
- Gene Name
- low density lipoprotein receptor class A domain containing 2
View Full Product Details