Product Details

SNP ID
rs7969189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8456680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTGCATGTTTATGGGCTCCATGT[C/G]ATATTTTGACACAAGAATATGATGC
Phenotype
MIM: 613579
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC6A PubMed Links

Gene Details

Gene
CLEC6A
Gene Name
C-type lectin domain family 6 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007033.1 Intron NP_001007034.1
NM_001317999.1 Intron NP_001304928.1

View Full Product Details