Product Details

SNP ID
rs13375457
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:93874471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAACAGAGGGGTTGTGAGAAG[G/T]ACTGGAGGTAAAATATTCCCCAAGG
Phenotype
MIM: 611199
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014597.4 Intron NP_055412.2

View Full Product Details