Product Details

SNP ID
hCV30761150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87870296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTTTCAAGTTATAAGTGTTTT[A/T]AAAAATAAGGGTATTCCTTTAGGAA
Phenotype
MIM: 612105 MIM: 601728
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLLN PubMed Links

Gene Details

Gene
KLLN
Gene Name
killin, p53-regulated DNA replication inhibitor
There are no transcripts associated with this gene.

Gene
PTEN
Gene Name
phosphatase and tensin homolog
There are no transcripts associated with this gene.

View Full Product Details