Product Details

SNP ID
rs12241664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88177886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGGTTGTTTGTGGTGGCTGTGG[A/C]ATGGTTTTGTTGAGGATGTGAACAT
Phenotype
MIM: 609360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNLS PubMed Links

Gene Details

Gene
RNLS
Gene Name
renalase, FAD dependent amine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031709.2 6716 Intron NP_001026879.2
NM_018363.3 6716 Intron NP_060833.1
XM_005269946.1 6716 Intron XP_005270003.1
XM_005269947.1 6716 Intron XP_005270004.1
XM_005269948.2 6716 Intron XP_005270005.1
XM_005269949.4 6716 Intron XP_005270006.1
XM_005269950.3 6716 Intron XP_005270007.1
XM_011539924.2 6716 UTR 3 XP_011538226.1
XM_011539927.2 6716 Intron XP_011538229.1
XM_017016380.1 6716 Intron XP_016871869.1
XM_017016381.1 6716 Intron XP_016871870.1
XM_017016382.1 6716 UTR 3 XP_016871871.1
XM_017016383.1 6716 Intron XP_016871872.1
XM_017016384.1 6716 Intron XP_016871873.1
XM_017016385.1 6716 Intron XP_016871874.1

View Full Product Details