Product Details

SNP ID
rs11634547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48669947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATATATTACAGTATACTCTATA[G/T]TTCCTGGCTCAAGGAGCTTAGGATT
Phenotype
MIM: 613529
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP152 PubMed Links

Gene Details

Gene
CEP152
Gene Name
centrosomal protein 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194998.1 Intron NP_001181927.1
NM_014985.3 Intron NP_055800.2
XM_006720437.3 Intron XP_006720500.1
XM_011521373.2 Intron XP_011519675.1
XM_011521374.2 Intron XP_011519676.1
XM_011521375.2 Intron XP_011519677.1
XM_011521378.2 Intron XP_011519680.1
XM_011521379.2 Intron XP_011519681.1
XM_011521381.2 Intron XP_011519683.1
XM_017022014.1 Intron XP_016877503.1
XM_017022015.1 Intron XP_016877504.1
XM_017022016.1 Intron XP_016877505.1

View Full Product Details