Product Details

SNP ID
rs12491394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51678153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTCCTGACTAGAAAAACTGCT[C/T]TGCCTTTAGCCCTGCTTCTGCGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD54L2 PubMed Links

Gene Details

Gene
RAD54L2
Gene Name
RAD54-like 2 (S. cerevisiae)
There are no transcripts associated with this gene.

Gene
TEX264
Gene Name
testis expressed 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129884.2 Intron NP_001123356.1
NM_001243725.1 Intron NP_001230654.1
NM_001243726.1 Intron NP_001230655.1
NM_001243727.2 Intron NP_001230656.1
NM_001278195.1 Intron NP_001265124.1
NM_015926.5 Intron NP_057010.1
XM_006713195.3 Intron XP_006713258.1
XM_006713197.3 Intron XP_006713260.1
XM_006713198.2 Intron XP_006713261.1
XM_011533805.2 Intron XP_011532107.1
XM_017006573.1 Intron XP_016862062.1
XM_017006574.1 Intron XP_016862063.1

View Full Product Details