Product Details

SNP ID
rs7010488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24386800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTAAATTTTCTCTCTCCTGCCG[G/T]GTCTGTCCCGCAGACCCTGGCTGAC
Phenotype
MIM: 606393
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMDEC1 PubMed Links

Gene Details

Gene
ADAMDEC1
Gene Name
ADAM like decysin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145271.1 Intron NP_001138743.1
NM_001145272.1 Intron NP_001138744.1
NM_014479.3 Intron NP_055294.1
Gene
LOC101929294
Gene Name
uncharacterized LOC101929294
There are no transcripts associated with this gene.

View Full Product Details