Product Details
- SNP ID
-
rs11259096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:14436619 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTGGTCAAGTGTTGTTAAGGCATG[C/T]TCAGGAACAATACTTGACCACATTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1265
PubMed Links
Gene Details
- Gene
- MIR1265
- Gene Name
- microRNA 1265
There are no transcripts associated with this gene.
View Full Product Details