Product Details

SNP ID
rs11114751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:81080279 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTTGGAAGTGAGTGTCAATGTC[C/G]CAGCAAGTAGAGCTTAAGACCAGGA
Phenotype
MIM: 614356
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACSS3 PubMed Links

Gene Details

Gene
ACSS3
Gene Name
acyl-CoA synthetase short-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269150.2 Intron XP_005269207.1
XM_005269151.4 Intron XP_005269208.1
XM_011538740.2 Intron XP_011537042.1

View Full Product Details