Product Details

SNP ID
rs13418293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98354428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAAATGTCTATTCAAATCCTTTG[A/C]CTGCTTTTTAATTGAGTTATTTGAC
Phenotype
MIM: 600053
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNGA3 PubMed Links

Gene Details

Gene
CNGA3
Gene Name
cyclic nucleotide gated channel alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079878.1 Intron NP_001073347.1
NM_001298.2 Intron NP_001289.1
XM_006712243.2 Intron XP_006712306.1
XM_011510554.2 Intron XP_011508856.1

View Full Product Details