Product Details

SNP ID
rs10994864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:61664701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACACGCTAATTCGCTGCTTGGTG[A/G]AAACTCCATTTTCCGTTTTAATGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf107 PubMed Links

Gene Details

Gene
C10orf107
Gene Name
chromosome 10 open reading frame 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173554.2 Intron NP_775825.1
XM_005269598.3 Intron XP_005269655.1
XM_005269599.4 Intron XP_005269656.1
XM_005269600.4 Intron XP_005269657.1
XM_006717690.3 Intron XP_006717753.1
XM_006717692.3 Intron XP_006717755.1

View Full Product Details