Product Details
- SNP ID
-
rs10985085
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:120903019 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTTTCTCATGGGGGCTTGTTCTA[C/T]AAATCCCCCCTTGGGCTCAACACTC
- Phenotype
-
MIM: 609740
MIM: 601711
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PHF19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7037154] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHF19
- Gene Name
- PHD finger protein 19
- Gene
- TRAF1
- Gene Name
- TNF receptor associated factor 1
View Full Product Details