Product Details

SNP ID
rs12151277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58064291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTTACCCAATATATTAGCTTC[C/G]TATTGCTCCTCCTGCAACCAGTTAC
Phenotype
MIM: 604077
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF135 PubMed Links

Gene Details

Gene
ZNF135
Gene Name
zinc finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164527.1 Intron NP_001157999.1
NM_001164529.1 Intron NP_001158001.1
NM_001164530.1 Intron NP_001158002.1
NM_001289401.1 Intron NP_001276330.1
NM_001289402.1 Intron NP_001276331.1
NM_003436.3 Intron NP_003427.3
NM_007134.1 Intron NP_009065.1
XM_005259211.1 Intron XP_005259268.1
XM_006723362.3 Intron XP_006723425.1
XM_006723363.3 Intron XP_006723426.1
XM_017027240.1 Intron XP_016882729.1
XM_017027241.1 Intron XP_016882730.1
XM_017027242.1 Intron XP_016882731.1
Gene
ZSCAN1
Gene Name
zinc finger and SCAN domain containing 1
There are no transcripts associated with this gene.

View Full Product Details