Product Details

SNP ID
rs10859549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93683504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCTGTTGGAATGGCTGTCCCTA[A/G]GCTGACACTTGCTTCAACCTTCTCT
Phenotype
MIM: 603454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRADD PubMed Links

Gene Details

Gene
CRADD
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320099.1 Intron NP_001307028.1
NM_001320100.1 Intron NP_001307029.1
NM_001320101.1 Intron NP_001307030.1
NM_003805.4 Intron NP_003796.1
XM_017020142.1 Intron XP_016875631.1
XM_017020143.1 Intron XP_016875632.1
XM_017020144.1 Intron XP_016875633.1
XM_017020145.1 Intron XP_016875634.1

View Full Product Details