Product Details

SNP ID
rs13430695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:100470833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCTGAACATGTGTCGCTAAAAC[C/T]GAAGGGAAAGGCACTTCCTGGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMS PubMed Links

Gene Details

Gene
NMS
Gene Name
neuromedin S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011717.1 Intron NP_001011717.1

View Full Product Details