Product Details

SNP ID
rs11119748
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:211478843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCGGTGCCGCTCTACGACCTAA[C/G]TCAGCTTTGTGGCCAGAGGAAGAGG
Phenotype
MIM: 180040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RD3 PubMed Links

Gene Details

Gene
RD3
Gene Name
retinal degeneration 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164688.1 1312 UTR 3 NP_001158160.1
NM_183059.2 1312 UTR 3 NP_898882.1
XM_017001151.1 1312 UTR 3 XP_016856640.1

View Full Product Details