Product Details

SNP ID
rs12140372
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:237881886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTAGGAAACTACTTGTCAGTTC[C/T]GGCTCAAAACTAAGTCTCTGATGAT
Phenotype
MIM: 613514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130331 PubMed Links

Gene Details

Gene
LOC100130331
Gene Name
POTE ankyrin domain family, member F pseudogene
There are no transcripts associated with this gene.

Gene
ZP4
Gene Name
zona pellucida glycoprotein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021186.4 Intron NP_067009.1

View Full Product Details