Product Details

SNP ID
rs11000465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72945334 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCTTACTCTCCCTCTCTCTCAC[C/T]GTTCACTCTAGGGGAAGTTATGTCA
Phenotype
MIM: 611653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G12B PubMed Links

Gene Details

Gene
PLA2G12B
Gene Name
phospholipase A2 group XIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318124.1 Intron NP_001305053.1
NM_001318125.1 Intron NP_001305054.1
NM_032562.4 Intron NP_115951.2
XM_005270244.4 Intron XP_005270301.1

View Full Product Details