Product Details

SNP ID
rs12256374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72938371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCAATTAGGCAAGAGAAAAAAT[C/T]TAAAAAATCCAAATTGGAAAGGAAG
Phenotype
MIM: 609330 MIM: 611653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OIT3 PubMed Links

Gene Details

Gene
OIT3
Gene Name
oncoprotein induced transcript 3
There are no transcripts associated with this gene.

Gene
PLA2G12B
Gene Name
phospholipase A2 group XIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318124.1 Intron NP_001305053.1
NM_001318125.1 Intron NP_001305054.1
NM_032562.4 Intron NP_115951.2
XM_005270244.4 Intron XP_005270301.1

View Full Product Details