Product Details

SNP ID
rs3760065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57697942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCATCTCCAGACACCCAGAGAC[A/G]CTCCAGAATGGAGGTCCTGAGGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRG3 PubMed Links

Gene Details

Gene
ADGRG3
Gene Name
adhesion G protein-coupled receptor G3
There are no transcripts associated with this gene.

Gene
DRC7
Gene Name
dynein regulatory complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289162.1 82 UTR 5 NP_001276091.1
NM_001289163.1 82 UTR 5 NP_001276092.1
NM_032269.5 82 UTR 5 NP_115645.4
XM_011523377.1 82 UTR 5 XP_011521679.1
XM_011523378.1 82 UTR 5 XP_011521680.1

View Full Product Details