Product Details

SNP ID
rs6881831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172331477 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGGAAGCGTGAGTTGCAAGACGC[C/T]GAAGAATCTCATGTGGCCCAAATAA
Phenotype
MIM: 613293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3PXD2B PubMed Links

Gene Details

Gene
SH3PXD2B
Gene Name
SH3 and PX domains 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017995.2 Intron NP_001017995.1
NM_001308175.1 Intron NP_001295104.1
XM_017009351.1 Intron XP_016864840.1

View Full Product Details