Product Details

SNP ID
rs10878128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64270017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTGATGTTTCACTAGTCTTTTC[A/T]GGCCTGGGAATATACTTCAAATGGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf56 PubMed Links

Gene Details

Gene
C12orf56
Gene Name
chromosome 12 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099676.2 Intron NP_001093146.1
NM_001170633.1 Intron NP_001164104.1
XM_017018770.1 Intron XP_016874259.1

View Full Product Details