Product Details
- SNP ID
-
rs11101001
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:49160294 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACTCCCTTAACCAAGCAAACGAAG[A/C]ATCACCAACAGTGATGCAATACAAC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf128
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79222178] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C10orf128
- Gene Name
- chromosome 10 open reading frame 128
- Gene
- FAM170B-AS1
- Gene Name
- FAM170B antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details