Product Details

SNP ID
rs6900904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:106189704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACAGTACAAAGGAAACACCTGA[C/T]ACTGTTAAAGTAAACAATATTTTTG
Phenotype
MIM: 604261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG5 PubMed Links

Gene Details

Gene
ATG5
Gene Name
autophagy related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286106.1 Intron NP_001273035.1
NM_001286107.1 Intron NP_001273036.1
NM_001286108.1 Intron NP_001273037.1
NM_001286111.1 Intron NP_001273040.1
NM_004849.3 Intron NP_004840.1

View Full Product Details