Product Details

SNP ID
rs11625771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74320996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAGGCTGTGGATAAGGATTGA[A/G]AATTGCCCACTGGAAATGCATATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VRTN PubMed Links

Gene Details

Gene
VRTN
Gene Name
vertebrae development associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018228.2 Intron NP_060698.2
XM_011536911.1 Intron XP_011535213.1

View Full Product Details