Product Details

SNP ID
rs12035175
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:12067281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAACATCATGTTCCTTGCCAGTG[C/T]TAAGGCTAAACAAGCCTTCCTCCCA
Phenotype
MIM: 153243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF8 PubMed Links

Gene Details

Gene
TNFRSF8
Gene Name
TNF receptor superfamily member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243.4 Intron NP_001234.3
NM_001281430.2 Intron NP_001268359.2
XM_011542441.2 Intron XP_011540743.1
XM_011542443.2 Intron XP_011540745.1

View Full Product Details