Product Details

SNP ID
rs11101143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49515779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGATCATGTTTGGCTGCTGAACTT[C/G]TATCTTCTTTTTCAGTTTCTGGGAG
Phenotype
MIM: 609413
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 1592 Intron NP_000115.1
NM_001277058.1 1592 Missense Mutation CAA,GAA Q914E NP_001263987.1
NM_001277059.1 1592 Missense Mutation CAA,GAA Q914E NP_001263988.1
Gene
PGBD3
Gene Name
piggyBac transposable element derived 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170753.3 1592 Missense Mutation CAA,GAA Q446E NP_736609.2

View Full Product Details