Product Details

SNP ID
rs11212377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107794536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCTTTCAACTGGAAAAGCAAA[A/C]TCAACTTATGATTAATCCTGCTAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC35F2 PubMed Links

Gene Details

Gene
SLC35F2
Gene Name
solute carrier family 35 member F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017515.4 Intron NP_059985.2

View Full Product Details