Product Details

SNP ID
rs12292191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70276320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTTGTAGCCTGGGGAAGCAAC[C/T]TCAGGAGCTCCTGAGAAAGAGCACC
Phenotype
MIM: 611054
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR548K PubMed Links

Gene Details

Gene
MIR548K
Gene Name
microRNA 548k
There are no transcripts associated with this gene.

Gene
PPFIA1
Gene Name
PTPRF interacting protein alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003626.3 Intron NP_003617.1
NM_177423.2 Intron NP_803172.1
XM_006718716.3 Intron XP_006718779.1
XM_011545306.2 Intron XP_011543608.1
XM_011545307.2 Intron XP_011543609.1
XM_011545308.2 Intron XP_011543610.1
XM_011545309.2 Intron XP_011543611.1
XM_011545310.2 Intron XP_011543612.1
XM_011545311.2 Intron XP_011543613.1
XM_011545312.2 Intron XP_011543614.1
XM_011545313.2 Intron XP_011543615.1
XM_011545314.2 Intron XP_011543616.1
XM_011545315.2 Intron XP_011543617.1
XM_011545316.2 Intron XP_011543618.1
XM_011545317.2 Intron XP_011543619.1
XM_011545318.2 Intron XP_011543620.1
XM_011545319.1 Intron XP_011543621.1
XM_017018448.1 Intron XP_016873937.1
XM_017018449.1 Intron XP_016873938.1
XM_017018450.1 Intron XP_016873939.1

View Full Product Details