Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195194.1 | 5025 | Missense Mutation | CCA,TCA | P101S | NP_001182123.1 |
NM_001195195.1 | 5025 | Intron | NP_001182124.1 | ||
NM_001251964.1 | 5025 | UTR 3 | NP_001238893.1 | ||
NM_022112.2 | 5025 | Missense Mutation | CCA,TCA | P105S | NP_071395.2 |
XM_017018113.1 | 5025 | Missense Mutation | CCA,TCA | P127S | XP_016873602.1 |
XM_017018114.1 | 5025 | Missense Mutation | CCA,TCA | P123S | XP_016873603.1 |
XM_017018115.1 | 5025 | Intron | XP_016873604.1 | ||
XM_017018116.1 | 5025 | Missense Mutation | CCA,TCA | P105S | XP_016873605.1 |
XM_017018117.1 | 5025 | Intron | XP_016873606.1 |