Product Details
- SNP ID
-
rs10752642
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
66
- Location
-
Chr.1:154509148 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATGCAAAATATTGTGCCTGCAGT[C/T]TGGCATGCTTAAAGATAAACAACCT
- Phenotype
-
MIM: 610482
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SHE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59771172] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHE
- Gene Name
- Src homology 2 domain containing E
There are no transcripts associated with this gene.
- Gene
- TDRD10
- Gene Name
- tudor domain containing 10
View Full Product Details