Product Details

SNP ID
rs11673599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42327224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTGTGAAGTACTTCAATAAAA[A/G]CATTTTTCTTCAGTCATTCATGTGC
Phenotype
MIM: 604267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEGF8 PubMed Links

Gene Details

Gene
MEGF8
Gene Name
multiple EGF like domains 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271938.1 Intron NP_001258867.1
NM_001410.2 Intron NP_001401.2
Gene
TMEM145
Gene Name
transmembrane protein 145
There are no transcripts associated with this gene.

View Full Product Details