Product Details

SNP ID
rs12320646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50924335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCTCATGGCCTGAAACAATCT[C/T]TCAAGTTAAATTTTAAAAGCGCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL7A PubMed Links

Gene Details

Gene
METTL7A
Gene Name
methyltransferase like 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014033.3 Intron NP_054752.3
XM_006719332.1 Intron XP_006719395.1

View Full Product Details