Product Details

SNP ID
rs10749467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122386196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAGACATTTTAGTAGGTATGTTA[C/T]AGTATCTTGTGGTTTTTATTTGCAT
Phenotype
MIM: 607772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHA1 PubMed Links

Gene Details

Gene
PLEKHA1
Gene Name
pleckstrin homology domain containing A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001974.2 Intron NP_001001974.1
NM_001195608.1 Intron NP_001182537.1
NM_021622.4 Intron NP_067635.2
XM_005270016.1 Intron XP_005270073.1
XM_005270019.1 Intron XP_005270076.1
XM_005270020.1 Intron XP_005270077.1
XM_005270021.4 Intron XP_005270078.1
XM_011540017.1 Intron XP_011538319.1
XM_011540018.1 Intron XP_011538320.1
XM_011540019.1 Intron XP_011538321.1
XM_011540021.1 Intron XP_011538323.1
XM_011540022.1 Intron XP_011538324.1
XM_011540024.1 Intron XP_011538326.1
XM_017016478.1 Intron XP_016871967.1
XM_017016479.1 Intron XP_016871968.1
XM_017016480.1 Intron XP_016871969.1
XM_017016481.1 Intron XP_016871970.1
XM_017016482.1 Intron XP_016871971.1
XM_017016483.1 Intron XP_016871972.1
XM_017016484.1 Intron XP_016871973.1
XM_017016485.1 Intron XP_016871974.1
XM_017016486.1 Intron XP_016871975.1
XM_017016487.1 Intron XP_016871976.1
XM_017016488.1 Intron XP_016871977.1
XM_017016489.1 Intron XP_016871978.1
XM_017016490.1 Intron XP_016871979.1
XM_017016491.1 Intron XP_016871980.1

View Full Product Details