Product Details

SNP ID
rs13078246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:31661636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCTCCCTTATGGACAGTGATC[A/G]GGCCATGCCAGTTCACCTTCATTTT
Phenotype
MIM: 606738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL10 PubMed Links

Gene Details

Gene
OSBPL10
Gene Name
oxysterol binding protein like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174060.1 2086 UTR 3 NP_001167531.1
NM_017784.4 2086 UTR 3 NP_060254.2
XM_005264843.3 2086 Intron XP_005264900.1
XM_005264844.4 2086 Intron XP_005264901.1
XM_005264845.1 2086 UTR 3 XP_005264902.1
XM_011533326.2 2086 Intron XP_011531628.1
XM_017005667.1 2086 Intron XP_016861156.1
XM_017005668.1 2086 Intron XP_016861157.1
XM_017005669.1 2086 Intron XP_016861158.1
XM_017005670.1 2086 Intron XP_016861159.1

View Full Product Details