Product Details

SNP ID
rs11037390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43261827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGTGTTCACAAAAATTACATAG[A/G]GGAACATTTTTTTAAACCAGTAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPKP3 PubMed Links

Gene Details

Gene
HNRNPKP3
Gene Name
heterogeneous nuclear ribonucleoprotein K pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details