Product Details

SNP ID
rs11033541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36300599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATATCATCTCACCTTCTAGCTCC[C/T]AGGAGGTACTTAACCTAAAGATGAC
Phenotype
MIM: 611728
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRR5L PubMed Links

Gene Details

Gene
PRR5L
Gene Name
proline rich 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160167.1 Intron NP_001153639.1
NM_001160168.1 Intron NP_001153640.1
NM_001160169.1 Intron NP_001153641.1
NM_024841.4 Intron NP_079117.3

View Full Product Details