Product Details
- SNP ID
-
rs11805141
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
20
- Location
-
Chr.1:5864679 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAACCAACCCTGACATGACTGTGGC[C/T]GCGACTTGGGTCCACACCAGGCAAG
- Phenotype
-
MIM: 607215
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4689
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114555857] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4689
- Gene Name
- microRNA 4689
There are no transcripts associated with this gene.
- Gene
- NPHP4
- Gene Name
- nephronophthisis 4
View Full Product Details