Product Details

SNP ID
rs10897430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63508091 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCTGGGAAGGCCATTGTTAAGTG[C/T]CTCTCCAGGGGTCCATTTCAGAGTT
Phenotype
MIM: 606096
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS12 PubMed Links

Gene Details

Gene
LGALS12
Gene Name
galectin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142535.1 105 Intron NP_001136007.1
NM_001142536.1 105 Intron NP_001136008.1
NM_001142537.1 105 UTR 5 NP_001136009.1
NM_001142538.1 105 UTR 5 NP_001136010.1
NM_033101.3 105 Intron NP_149092.2
XM_017018458.1 105 Intron XP_016873947.1

View Full Product Details