Product Details
- SNP ID
-
rs12923774
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:14645866 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACACACATACACACACATACATA[C/T]ATTCATTGTTTATATTCTTTGAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BFAR
PubMed Links
Gene Details
- Gene
- BFAR
- Gene Name
- bifunctional apoptosis regulator
View Full Product Details