Product Details

SNP ID
rs12037104
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:211659753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTGGAAGGAATTCCAAGGCAGG[C/T]GAATGATACATTTTTTCCCCATCTC
Phenotype
MIM: 604043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105748977 PubMed Links

Gene Details

Gene
LOC105748977
Gene Name
uncharacterized LOC105748977
There are no transcripts associated with this gene.

Gene
NEK2
Gene Name
NIMA related kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204182.1 Intron NP_001191111.1
NM_001204183.1 Intron NP_001191112.1
NM_002497.3 Intron NP_002488.1
XM_005273147.1 Intron XP_005273204.1

View Full Product Details