Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002830.3 | Intron | NP_002821.1 | |||
XM_011511560.2 | Intron | XP_011509862.1 | |||
XM_011511562.2 | Intron | XP_011509864.1 | |||
XM_011511563.2 | Intron | XP_011509865.1 | |||
XM_017004596.1 | Intron | XP_016860085.1 | |||
XM_017004597.1 | Intron | XP_016860086.1 | |||
XM_017004598.1 | Intron | XP_016860087.1 | |||
XM_017004599.1 | Intron | XP_016860088.1 | |||
XM_017004600.1 | Intron | XP_016860089.1 | |||
XM_017004601.1 | Intron | XP_016860090.1 | |||
XM_017004602.1 | Intron | XP_016860091.1 | |||
XM_017004603.1 | Intron | XP_016860092.1 | |||
XM_017004604.1 | Intron | XP_016860093.1 | |||
XM_017004605.1 | Intron | XP_016860094.1 | |||
XM_017004606.1 | Intron | XP_016860095.1 | |||
XM_017004607.1 | Intron | XP_016860096.1 | |||
XM_017004608.1 | Intron | XP_016860097.1 |