Product Details

SNP ID
rs11149808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75219863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCTCTCTGGAATGTGAGCTTG[A/T]TTCTGAACTTTCTATGAATAATTGC
Phenotype
MIM: 602941 MIM: 118890
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAR1 PubMed Links

Gene Details

Gene
BCAR1
Gene Name
BCAR1, Cas family scaffolding protein
There are no transcripts associated with this gene.

Gene
CTRB1
Gene Name
chymotrypsinogen B1
There are no transcripts associated with this gene.

Gene
LOC100506281
Gene Name
uncharacterized LOC100506281
There are no transcripts associated with this gene.

View Full Product Details