Product Details
- SNP ID
-
rs11864719
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75221231 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTGTTGGGTTGGCCCGCGCACAG[G/T]AGTCGAGAATCTGGTCCTGACATAG
- Phenotype
-
MIM: 602941
MIM: 118890
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BCAR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139332712] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCAR1
- Gene Name
- BCAR1, Cas family scaffolding protein
There are no transcripts associated with this gene.
- Gene
- CTRB1
- Gene Name
- chymotrypsinogen B1
There are no transcripts associated with this gene.
- Gene
- LOC100506281
- Gene Name
- uncharacterized LOC100506281
There are no transcripts associated with this gene.
View Full Product Details