Product Details

SNP ID
rs12830141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52567830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACGAAACTGACACTATCGCCTTC[A/T]TCTTACAGATGAGAAAACAAGAGTC
Phenotype
MIM: 608248
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT74 PubMed Links

Gene Details

Gene
KRT74
Gene Name
keratin 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175053.3 Intron NP_778223.2

View Full Product Details