Product Details

SNP ID
rs12578695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55686984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTACCTCATCCCCTTACATCCT[A/C]ACCCTACTACCCCACCTTCCTCTGA
Phenotype
MIM: 600536
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144996.1 Intron NP_001138468.1
NM_001144997.1 Intron NP_001138469.1
NM_002206.2 Intron NP_002197.2
XM_005268839.1 Intron XP_005268896.1
XM_005268840.1 Intron XP_005268897.1
XM_005268841.1 Intron XP_005268898.1
XM_005268842.1 Intron XP_005268899.1
XM_005268844.1 Intron XP_005268901.1
XM_005268846.1 Intron XP_005268903.1
XM_005268848.1 Intron XP_005268905.1
XM_005268849.1 Intron XP_005268906.1
XM_005268850.1 Intron XP_005268907.1
XM_017019265.1 Intron XP_016874754.1
Gene
METTL7B
Gene Name
methyltransferase like 7B
There are no transcripts associated with this gene.

View Full Product Details