Product Details

SNP ID
rs10748185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71942075 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAGTAGCAAGAGCCTGCAACTT[A/G]ATTTGATTTGTTCAAATTTGAATTT
Phenotype
MIM: 607478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPH2 PubMed Links

Gene Details

Gene
TPH2
Gene Name
tryptophan hydroxylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173353.3 Intron NP_775489.2

View Full Product Details