Product Details

SNP ID
rs7671215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145768665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCAGTTCATTAAGCAATCTAGT[A/G]ACTGTAAATGTTAGAACCTAACTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf51 PubMed Links

Gene Details

Gene
C4orf51
Gene Name
chromosome 4 open reading frame 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080531.1 Intron NP_001074000.1
XM_006714289.3 Intron XP_006714352.1
XM_011532196.2 Intron XP_011530498.1
XM_011532197.2 Intron XP_011530499.1
XM_017008547.1 Intron XP_016864036.1
XM_017008548.1 Intron XP_016864037.1
XM_017008549.1 Intron XP_016864038.1
XM_017008550.1 Intron XP_016864039.1
XM_017008551.1 Intron XP_016864040.1
XM_017008552.1 Intron XP_016864041.1
XM_017008553.1 Intron XP_016864042.1
XM_017008554.1 Intron XP_016864043.1
XM_017008555.1 Intron XP_016864044.1
XM_017008556.1 Intron XP_016864045.1
Gene
ZNF827
Gene Name
zinc finger protein 827
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306215.1 Intron NP_001293144.1
NM_178835.4 Intron NP_849157.2
XM_011531634.2 Intron XP_011529936.1
XM_011531635.2 Intron XP_011529937.1
XM_011531645.2 Intron XP_011529947.1
XM_017007768.1 Intron XP_016863257.1
XM_017007769.1 Intron XP_016863258.1
XM_017007770.1 Intron XP_016863259.1
XM_017007771.1 Intron XP_016863260.1
XM_017007772.1 Intron XP_016863261.1
XM_017007773.1 Intron XP_016863262.1
XM_017007774.1 Intron XP_016863263.1
XM_017007775.1 Intron XP_016863264.1
XM_017007776.1 Intron XP_016863265.1
XM_017007777.1 Intron XP_016863266.1

View Full Product Details