Product Details
- SNP ID
-
rs6536991
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:140560427 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCTTCACTCCCAAAACATGTCTT[C/T]TCTTCACTGACATGAACTTTTTCAA
- Phenotype
-
MIM: 610196
MIM: 113730
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ELMOD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78724285] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ELMOD2
- Gene Name
- ELMO domain containing 2
There are no transcripts associated with this gene.
- Gene
- UCP1
- Gene Name
- uncoupling protein 1
View Full Product Details